Assay Details
Gene Symbol: MIR543 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_033040.1 | 1 | 35 | 61 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA mir-1193 | MIR1193 | eca-mir-1193 | -1.6kb | Sense | Chr. 24 - 43341207 - 43341327 on NCBI Build EquCab3.0 |
Downstream | microRNA mir-495 | MIR495 | eca-mir-495 | 1.8kb | Sense | Chr. 24 - 43344716 - 43344797 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
