Assay Details
Gene Symbol: MIR770 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_033045.1 | 1 | 52 | 64 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | delta like non-canonical Notch ligand 1 | DLK1 | -100.1kb | Sense | Chr. 24 - 43042141 - 43050221 on NCBI Build EquCab3.0 | |
Downstream | microRNA mir-493b | MIR493B | eca-mir-493b | 17.2kb | Sense | Chr. 24 - 43167633 - 43167721 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
