Assay Details
Gene Symbol: MIR181A-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_033064.1 | 1 | 57 | 64 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | protein tyrosine phosphatase receptor type C | PTPRC | -98.9kb | Anti-sense | Chr. 30 - 27521147 - 27635419 on NCBI Build EquCab3.0 | |
Downstream | nuclear receptor subfamily 5 group A member 2 | NR5A2 | SF-2 | 902.6kb | Anti-sense | Chr. 30 - 28637061 - 28769094 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
