Assay Details
Gene Symbol: MIR194-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_033065.1 | 1 | 43 | 73 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA mir-215 | MIR215 | eca-mir-215 | -195.0 | Sense | Chr. 30 - 10715793 - 10715902 on NCBI Build EquCab3.0 |
Downstream | RAB3 GTPase activating non-catalytic protein subunit 2 | RAB3GAP2 | 34.6kb | Sense | Chr. 30 - 10750767 - 10849278 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
