Assay Details
Gene Symbol: MIR215 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_033066.1 | 1 | 50 | 83 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
3'(2'), 5'-bisphosphate nucleotidase 1 |
BPNT1 | -34.4kb | Sense | Chr. 30 - 10660396 - 10681381 on NCBI Build EquCab3.0 | |
Downstream | microRNA mir-194-2 | MIR194-2 | eca-mir-194-2 | 195.0 | Sense | Chr. 30 - 10716097 - 10716181 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
