Assay Details
Gene Symbol: MIR1468 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_033073.1 | 1 | 50 | 71 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | spindlin family member 4 | SPIN4 | -658.9kb | Sense | Chr. X - 48871191 - 48879776 on NCBI Build EquCab3.0 | |
Downstream | APC membrane recruitment protein 1 | AMER1 | 382.4kb | Sense | Chr. X - 49921193 - 49942678 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
