Assay Details
Gene Symbol: MIR19B-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_033077.1 | 1 | 41 | 72 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA mir-92a-2 | MIR92A-2 | eca-mir-92a-2 | -57.0 | Sense | Chr. X - 110758524 - 110758598 on NCBI Build EquCab3.0 |
Downstream | microRNA mir-20b | MIR20B | eca-mir-20b | 43.0 | Sense | Chr. X - 110758793 - 110758861 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
