Assay Details
Gene Symbol: MIR221 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_033079.1 | 1 | 57 | 72 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | divergent protein kinase domain 2B | DIPK2B | CXHXorf36 | -416.5kb | Sense | Chr. X - 38052758 - 38097349 on NCBI Build EquCab3.0 |
Downstream | microRNA mir-222 | MIR222 | eca-mir-222 | 670.0 | Sense | Chr. X - 38514675 - 38514784 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
