Assay Details
Gene Symbol: MIR222 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_033080.1 | 1 | 48 | 68 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA mir-221 | MIR221 | eca-mir-221 | -670.0 | Sense | Chr. X - 38513896 - 38514005 on NCBI Build EquCab3.0 |
Downstream | zinc finger protein 674 | LOC100060244 | 553.8kb | Anti-sense | Chr. X - 39068567 - 39081774 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
