Assay Details
Gene Symbol: MIR223 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_033081.1 | 1 | 51 | 95 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | moesin | MSN | -292.5kb | Sense | Chr. X - 51233198 - 51295218 on NCBI Build EquCab3.0 | |
Downstream | V-set and immunoglobulin domain containing 4 | VSIG4 | 2.7kb | Anti-sense | Chr. X - 51590592 - 51615203 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
