Assay Details
Gene Symbol: MIR374B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_033088.1 | 1 | 35 | 72 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA mir-421 | MIR421 | eca-mir-421 | -87.0 | Sense | Chr. X - 58556025 - 58556103 on NCBI Build EquCab3.0 |
Downstream | microRNA mir-545 | MIR545 | eca-mir-545 | 64.3kb | Sense | Chr. X - 58620584 - 58620689 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
