Assay Details
Gene Symbol: MIR542 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_033108.1 | 1 | 45 | 86 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA mir-450c | MIR450C |
MIR451A, eca-mir-450c, eca-mir-451a |
-700.0 | Sense | Chr. X - 111015523 - 111015611 on NCBI Build EquCab3.0 |
Downstream | microRNA mir-503 | MIR503 | eca-mir-503 | 4.7kb | Sense | Chr. X - 111021114 - 111021184 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
