Assay Details
Gene Symbol: RN28S | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_046309.3 | 1 | 322 | 109 | ||
GenBank mRNA | EU081775.1 | - | 1 | 322 | 109 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | surfactant protein D | SFTPD | -123.3kb | Sense | Chr. 1 - 89805789 - 89817016 on NCBI Build EquCab3.0 | |
Downstream | uncharacterized LOC100066811 | LOC100066811 | FAM154B | 100.6kb | Sense | Chr. 1 - 90041661 - 90050003 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
