Assay Details
Gene Symbol: MIR8918 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_127904.1 | 1 | 57 | 71 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
myomixer, myoblast fusion factor |
MYMX | -12.1kb | Anti-sense | Chr. 20 - 43984692 - 43989315 on NCBI Build EquCab3.0 | |
Downstream | heat shock protein 90 alpha family class B member 1 | HSP90AB1 | HSP90 | 13.9kb | Anti-sense | Chr. 20 - 44015470 - 44021393 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
