Assay Details
Gene Symbol: MIR8965 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_127969.1 | 1 | 54 | 71 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | erythrocyte membrane protein band 4.1 like 1 | EPB41L1 | -20.5kb | Sense | Chr. 22 - 27597108 - 27715259 on NCBI Build EquCab3.0 | |
Downstream | DLG associated protein 4 | DLGAP4 | 43.7kb | Sense | Chr. 22 - 27779621 - 27975759 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
