Assay Details
Gene Symbol: MIR9011 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128023.1 | 1 | 121 | 69 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | PH domain containing endocytic trafficking adaptor 1 | PHETA1 | FAM109A | -207.5kb | Anti-sense | Chr. 8 - 23213671 - 23220294 on NCBI Build EquCab3.0 |
Downstream | myosin light chain 2 | MYL2 | 121.1kb | Anti-sense | Chr. 8 - 23549109 - 23557228 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
