Assay Details
Gene Symbol: MIR9016 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128028.1 | 1 | 124 | 65 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | eukaryotic translation elongation factor 1 delta | EEF1D | -4.4kb | Anti-sense | Chr. 9 - 84403650 - 84419240 on NCBI Build EquCab3.0 | |
Downstream | pyrroline-5-carboxylate reductase 3 | PYCR3 | PYCRL | 1.7kb | Anti-sense | Chr. 9 - 84425538 - 84430923 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
