Assay Details
Gene Symbol: MIR9017 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128030.1 | 1 | 116 | 59 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | zinc finger CCCH-type containing 7B | ZC3H7B | -1.9kb | Anti-sense | Chr. 28 - 38932158 - 38985559 on NCBI Build EquCab3.0 | |
Downstream |
TEF transcription factor, PAR bZIP family member |
TEF | 2.3kb | Anti-sense | Chr. 28 - 38989853 - 39013715 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
