Assay Details
Gene Symbol: MIR9026 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128040.1 | 1 | 120 | 89 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | protein O-fucosyltransferase 2 | POFUT2 | -150.7kb | Anti-sense | Chr. 26 - 41665510 - 41680747 on NCBI Build EquCab3.0 | |
Downstream | solute carrier family 19 member 1 | SLC19A1 | 88.4kb | Anti-sense | Chr. 26 - 41919982 - 41943814 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
