Assay Details
Gene Symbol: MIR9037 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128051.1 | 1 | 23 | 52 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | adhesion G protein-coupled receptor D2 | ADGRD2 | GPR144 | Anti-sense | Chr. 25 - 29048954 - 29075694 on NCBI Build EquCab3.0 | |
Downstream | nuclear receptor subfamily 5 group A member 1 | NR5A1 | SF-1 | 9.1kb | Sense | Chr. 25 - 29079410 - 29103794 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
