Assay Details
Gene Symbol: MIR490 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032828.1 | 1 | 40 | 58 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | myotrophin | MTPN | -672.2kb | Anti-sense | Chr. 4 - 90157144 - 90214361 on NCBI Build EquCab3.0 | |
Downstream | pleiotrophin | PTN | 232.1kb | Anti-sense | Chr. 4 - 91118734 - 91220261 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
