Assay Details
Gene Symbol: MIR592 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032829.1 | 1 | 42 | 62 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | glutamate metabotropic receptor 8 | GRM8 | Sense | Chr. 4 - 81973863 - 82689994 on NCBI Build EquCab3.0 | ||
Downstream | uncharacterized LOC102150275 | LOC102150275 | 188.9kb | Sense | Chr. 4 - 82697975 - 82727818 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
