Assay Details
Gene Symbol: MIR16-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032837.1 | 1 | 33 | 78 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | structural maintenance of chromosomes 4 | SMC4 | Sense | Chr. 19 - 3377517 - 3412413 on NCBI Build EquCab3.0 | ||
Downstream | microRNA mir-15b | MIR15B | eca-mir-15b | 78.0 | Sense | Chr. 19 - 3408021 - 3408084 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
