Assay Details
Gene Symbol: MIR200C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032856.1 | 1 | 33 | 62 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | protein tyrosine phosphatase non-receptor type 6 | PTPN6 | -2.6kb | Sense | Chr. 6 - 35420189 - 35436037 on NCBI Build EquCab3.0 | |
Downstream | microRNA mir-141 | MIR141 | eca-mir-141 | 346.0 | Sense | Chr. 6 - 35439079 - 35439150 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
