Assay Details
Gene Symbol: MIR763 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032859.1 | 1 | 63 | 112 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | high mobility group AT-hook 2 | HMGA2 | Sense | Chr. 6 - 82552985 - 82681843 on NCBI Build EquCab3.0 | ||
Downstream |
LLP homolog, long-term synaptic facilitation factor |
LLPH | 227.4kb | Anti-sense | Chr. 6 - 82811710 - 82818771 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
