Assay Details
Gene Symbol: MIR9075 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128098.1 | 1 | 120 | 66 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | spermatogenesis-associated protein 2-like protein | LOC102147996 | -30.0kb | Anti-sense | Chr. 3 - 17803657 - 17812993 on NCBI Build EquCab3.0 | |
Downstream | chemokine like factor | CKLF | 9.7kb | Anti-sense | Chr. 3 - 17852827 - 17864035 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
