Assay Details
Gene Symbol: MIR744 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128124.1 | 1 | 45 | 58 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | mitogen-activated protein kinase kinase 4 | MAP2K4 | Sense | Chr. 11 - 54691894 - 54828149 on NCBI Build EquCab3.0 | ||
Downstream | myocardin | MYOCD | 499.6kb | Sense | Chr. 11 - 55252088 - 55346468 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
