Assay Details
Gene Symbol: H19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_027326.3 | 3 - 4 | 1504 | 59 | ||
GenBank mRNA | DN511008.1 | - | 3 - 4 | 320 | 59 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC106781503 | LOC106781503 | -180.0 | Anti-sense | Chr. 12 - 34334439 - 34334918 on NCBI Build EquCab3.0 | |
Downstream | microRNA mir-675 | MIR675 | eca-mir-675 | 521.0 | Sense | Chr. 12 - 34335950 - 34336022 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
