Assay Details
Gene Symbol: MIR218-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032938.1 | 1 | 32 | 62 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | slit guidance ligand 3 | SLIT3 | Sense | Chr. 14 - 10902364 - 11489820 on NCBI Build EquCab3.0 | ||
Downstream | pantothenate kinase 3 | PANK3 | 190.6kb | Sense | Chr. 14 - 11586765 - 11611512 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
