Assay Details
| Gene Symbol: MIR874 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_032941.1 | 1 | 33 | 51 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | heterogeneous nuclear ribonucleoprotein A0 | HNRNPA0 | -104.1kb | Sense | Chr. 14 - 37917496 - 37919311 on NCBI Build EquCab3.0 | |
| Downstream |
SPARC (osteonectin), cwcv and kazal like domains proteoglycan 1 |
SPOCK1 | 124.3kb | Sense | Chr. 14 - 38147763 - 38618573 on NCBI Build EquCab3.0 |
More Information
Set Membership: |
|

MIR874
Genomic Map