Assay Details
Gene Symbol: MIR711 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032955.1 | 1 | 36 | 61 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | ubiquinol-cytochrome c reductase core protein 1 | UQCRC1 | -20.5kb | Sense | Chr. 16 - 39920155 - 39929154 on NCBI Build EquCab3.0 | |
Downstream | 6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 4 | PFKFB4 | 17.8kb | Sense | Chr. 16 - 39967546 - 40010150 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
