Assay Details
Gene Symbol: MIR19A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032961.1 | 1 | 44 | 78 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | translation initiation factor IF-2-like | LOC111768661 | Sense | Chr. 17 - 61681419 - 61689144 on NCBI Build EquCab3.0 | ||
Downstream | microRNA mir-20a | MIR20A | eca-mir-20a | 90.0 | Sense | Chr. 17 - 61685489 - 61685559 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
