Assay Details
Gene Symbol: MIR19B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032962.1 | 1 | 31 | 63 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | translation initiation factor IF-2-like | LOC111768661 | Sense | Chr. 17 - 61681419 - 61689144 on NCBI Build EquCab3.0 | ||
Downstream | microRNA mir-92a | MIR92A | eca-mir-92a | 55.0 | Sense | Chr. 17 - 61685746 - 61685806 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
