Assay Details
Gene Symbol: MIR551B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032969.1 | 1 | 28 | 58 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | translation initiation factor IF-2-like | LOC111769059 | -413.0kb | Sense | Chr. 19 - 11132644 - 11134196 on NCBI Build EquCab3.0 | |
Downstream | MDS1 and EVI1 complex locus | MECOM | 432.4kb | Anti-sense | Chr. 19 - 11979645 - 12514266 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
