Assay Details
Gene Symbol: MIR9107 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128135.1 | 1 | 56 | 73 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | casein kinase 1 delta | CSNK1D | Sense | Chr. 11 - 947052 - 987043 on NCBI Build EquCab3.0 | ||
Downstream | microRNA mir-9098 | MIR9098 | eca-mir-9098 | 26.9kb | Sense | Chr. 11 - 981951 - 982085 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
