Assay Details
Gene Symbol: MIR9117 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128147.1 | 1 | 54 | 61 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | kinesin light chain 2 | KLC2 | Sense | Chr. 12 - 29980728 - 29991168 on NCBI Build EquCab3.0 | ||
Downstream |
RAB1B, member RAS oncogene family |
RAB1B | 8.5kb | Sense | Chr. 12 - 29991872 - 29999052 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
