Assay Details
Gene Symbol: MIR9138 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128171.1 | 1 | 116 | 97 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | translation initiation factor IF-2-like | LOC111767634 | -2.9kb | Anti-sense | Chr. 13 - 12359249 - 12364229 on NCBI Build EquCab3.0 | |
Downstream | neutrophil cytosolic factor 1 | NCF1 | 118.8kb | Sense | Chr. 13 - 12486118 - 12504022 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
