Assay Details
Gene Symbol: MIR9146 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128218.1 | 1 | 60 | 83 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA mir-1301 | MIR1301 | eca-mir-1301 | -104.8kb | Sense | Chr. 15 - 71646910 - 71646981 on NCBI Build EquCab3.0 |
Downstream | proopiomelanocortin | POMC |
ACTH, alpha-MSH |
27.0kb | Sense | Chr. 15 - 71778801 - 71784802 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
