Assay Details
Gene Symbol: MIR3613 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128239.1 | 1 | 21 | 109 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | tripartite motif containing 13 | TRIM13 | -175.0 | Anti-sense | Chr. 17 - 21074897 - 21087769 on NCBI Build EquCab3.0 | |
Downstream | SPRY domain containing 7 | SPRYD7 | 55.4kb | Sense | Chr. 17 - 21143459 - 21171127 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
