Assay Details
Gene Symbol: MIR9171 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128249.2 | 1 | 56 | 73 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | H1.10 linker histone | H1-10 | -725.0 | Sense | Chr. 16 - 2843762 - 2844885 on NCBI Build EquCab3.0 | |
Downstream | microRNA mir-9176 | MIR9176 | eca-mir-9176 | 2.7kb | Anti-sense | Chr. 16 - 2848408 - 2848552 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
