Assay Details
Gene Symbol: MIR684 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032815.1 | 1 | 47 | 82 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | bone morphogenetic protein receptor type 1B | BMPR1B | -145.5kb | Sense | Chr. 3 - 44399372 - 44776886 on NCBI Build EquCab3.0 | |
Downstream | hematopoietic prostaglandin D synthase | HPGDS | 250.4kb | Anti-sense | Chr. 3 - 45172821 - 45202617 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
