Assay Details
| Gene Symbol: MIR495 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033036.1 | 1 | 36 | 77 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA mir-543 | MIR543 | eca-mir-543 | -1.8kb | Sense | Chr. 24 - 43342877 - 43342948 on NCBI Build EquCab3.0 |
| Downstream | microRNA mir-3958 | MIR3958 | eca-mir-3958 | 1.9kb | Sense | Chr. 24 - 43346664 - 43346806 on NCBI Build EquCab3.0 |
More Information
Set Membership: |
|

MIR495
Genomic Map