Assay Details
Gene Symbol: MIR8954 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_127951.1 | 1 | 96 | 125 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | EMAP like 1 | EML1 | -22.6kb | Anti-sense | Chr. 24 - 42182478 - 42346474 on NCBI Build EquCab3.0 | |
Downstream | Enah/Vasp-like | EVL | 1.1kb | Anti-sense | Chr. 24 - 42370374 - 42530476 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
