Assay Details
Gene Symbol: MIR8964 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_127968.1 | 1 | 79 | 89 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | centrosomal protein 250 | CEP250 | Sense | Chr. 22 - 27098476 - 27141602 on NCBI Build EquCab3.0 | ||
Downstream | CMP-N-acetylneuraminate-beta-galactosamide-alpha-2,3-sialyltransferase 1 | LOC102148379 | 7.9kb | Anti-sense | Chr. 22 - 27148251 - 27150698 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
