Assay Details
Gene Symbol: MIR8966 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_127970.1 | 1 | 54 | 94 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | serine/threonine kinase 35 | STK35 | Sense | Chr. 22 - 20935700 - 21034610 on NCBI Build EquCab3.0 | ||
Downstream | prodynorphin | PDYN | 134.9kb | Anti-sense | Chr. 22 - 21126416 - 21141304 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
