Assay Details
Gene Symbol: MIR9001 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128011.1 | 1 | 63 | 71 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | A-kinase interacting protein 1 | AKIP1 | -9.2kb | Anti-sense | Chr. 7 - 80445884 - 80453351 on NCBI Build EquCab3.0 | |
Downstream | TMEM9 domain family member B | TMEM9B | 4.3kb | Sense | Chr. 7 - 80466975 - 80491662 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
