Assay Details
Gene Symbol: MIR9009 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128020.1 | 1 | 89 | 103 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | matrix metallopeptidase 17 | MMP17 | -38.9kb | Sense | Chr. 8 - 31552276 - 31581491 on NCBI Build EquCab3.0 | |
Downstream | pseudouridine synthase 1 | PUS1 | 5.1kb | Sense | Chr. 8 - 31625576 - 31639252 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
