Assay Details
Gene Symbol: MIR9020 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128033.1 | 1 | 90 | 108 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | ceramide kinase | CERK | -2.9kb | Anti-sense | Chr. 28 - 43632379 - 43696143 on NCBI Build EquCab3.0 | |
Downstream | TBC1 domain family member 22A | TBC1D22A | 24.7kb | Sense | Chr. 28 - 43723818 - 44066207 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
