Assay Details
| Gene Symbol: MIR1249 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_128035.1 | 1 | 81 | 111 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | KIAA0930 | KIAA0930 | Sense | Chr. 28 - 42324306 - 42365096 on NCBI Build EquCab3.0 | ||
| Downstream | uroplakin 3A | UPK3A | 62.5kb | Anti-sense | Chr. 28 - 42391658 - 42399591 on NCBI Build EquCab3.0 |
More Information
Set Membership: |
|

MIR1249
Genomic Map