Assay Details
Gene Symbol: MIR9022 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128036.1 | 1 | 74 | 99 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | tubulin tyrosine ligase like 8 | TTLL8 | -1.7kb | Sense | Chr. 28 - 46614092 - 46690367 on NCBI Build EquCab3.0 | |
Downstream | Mov10 like RISC complex RNA helicase 1 | MOV10L1 | 26.4kb | Anti-sense | Chr. 28 - 46718580 - 46787198 on NCBI Build EquCab3.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
